Order Entry
Canada
Orders LinkContactUsLinkComponent
PRC Pathway Modulator Set Abcam

PRC Pathway Modulator Set Abcam

PRC Pathway Modulator Set, 1 Kit
Catalog # ABCA_AB284177-1KIT
New Product
Supplier:   Abcam
Supplier #:  AB284177-1KIT
PRC Pathway Modulator Set Abcam
Catalog # ABCA_AB284177-1KIT
Supplier:  Abcam
New Product
CAS Number:  
PRC Pathway Modulator Set Abcam

PRC Pathway Modulator Set, 1 Kit

CAS Number:  
Price
$1,040.00
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PRC Pathway Modulator Set (ab284177) is part of the multiplex kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    PRC Pathway Modulator Set
  • Size:
    1 Kit
  • Cat. No.:
    ABCA_AB284177-1KIT
  • Storage temperature:
    Store at -20°C.

Specifications