Order Entry
Canada
Orders LinkContactUsLinkComponent
 

RIPA Buffer, Abcam

New Product
Supplier:   Abcam
RIPA Buffer, Abcam
Supplier:  Abcam
New Product
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.

  • Constituents: 1% Alcohol Ethoxylate, 0.877% Sodium chloride, 0.788% Tris HCl, 0.5% Sodium deoxycholate, 0.1% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCA_AB288006-500M
  • ABCA_AB288006-100M
  • Storage temperature:
  • Store at -20°C.
  • Store at -20°C.

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • RIPA Buffer
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $964.00
    $964.00
    Each (500ml)
       $964.00
     

    Supplier Number: AB288006-500ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB288006-500M
    Storage temperature
    Store at -20°C.
    New Product
  • RIPA Buffer
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $257.00
    $257.00
    Each (100ml)
       $257.00
     

    Supplier Number: AB288006-100ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB288006-100M
    Storage temperature
    Store at -20°C.
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.