AEC Substrate Buffer, Abcam
AEC Substrate Buffer, 50 ml
Catalog # ABCA_AB286857-50ML
New Product
Supplier: Abcam
: AB286857-50ML
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This product is manufactured by BioVision, an Abcam company and was previously called 2123 AEC Substrate Buffer.
- Constituents: 2.63% Sodium acetate, 0.3% Hydrogen peroxide
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:AEC Substrate Buffer
- Size:50 ml
- Cat. No.:ABCA_AB286857-50ML
- Storage temperature:Store at +4°C.



