Cloud Crystal Creation
Cloud Crystal Creation
Catalog # 470359-754
New Product
Supplier: THE GEM SHOP INC SE
Supplier #: GEM_SHOP_KT_1
Jump to:
Product Details & Documents
Teach students how crystals form in nature.
Study how crystals form in nature
Highly engaging
Practice observation and hypothesis formation
A chemical reaction occurs when this rock is combined with vinegar. Watch as beautiful detailed white crystals form on top of this rock as the vinegar evaporates. These crystal formations mimic formations formed in caves (stalactities and stalagmites), and encourage students to form observations, make a hypothesis, and discuss real world connections.
Specifications
- Description:Cloud Crystal Creation