Order Entry
Canada
Orders LinkContactUsLinkComponent
G-Biosciences TBE-Urea Sample Buffer | Avantor

G-Biosciences TBE-Urea Sample Buffer

TBE-Urea Sample Buffer, 30 mL
Catalog # CA95029-252
Supplier:   G-Biosciences
Fournisseur #:  786-474
G-Biosciences TBE-Urea Sample Buffer
Catalog # CA95029-252
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences TBE-Urea Sample Buffer

TBE-Urea Sample Buffer, 30 mL

CAS Number:  
Price
$125.84
Log in pour la disponibilité
Each (30ml)
Product Details & Documents

Loading buffer to resolve single-stranded DNA oligos or RNA into sharp, distinct bands.

  • Ready-to-use format: Pre-mixed buffer simplifies sample preparation and improves reproducibility

Broad application range: Used in molecular biology protocols involving RNA electrophoresis, ribonuclease protection assays, and sequencing gels.

Spécifications

Spécifications

  • Description:
    TBE-Urea Sample Buffer
  • Size:
    30 mL

Spécifications