G-Biosciences TAE Running Buffer (50X)
Catalog # CA82021-492
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:TAE, 50X
- Size:1 L
- Cat. no.:CA82021-492
- Supplier No.:R023
Specifications
About this item
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
50X Concentrated solution: 2 M Tris-Acetate, 0.05 M EDTA (pH 8.3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.