Order Entry
Canada
Orders LinkContactUsLinkComponent
 

G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

Supplier:   G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Supplier:  G-Biosciences
Certificates
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

2 Options Available Below

Product Details & Documents

Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

  • 1.5 M sodium chloride

  • 0.5 M  sodium hydroxide

A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • CA82021-476
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
    1 gallon
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $292.63
    $292.63
    Each (1l)
       $292.63
     

    Supplier Number: R015
  • CA82021-478
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
    1 L
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $459.06
    $459.06
    Each (4l)
       $459.06
     

    Supplier Number: R016