ContactUsLinkComponent
Multi-Analyte Photometer for Water Quality Analysis, V-3000
Multi-Analyte Photometer for Water Quality Analysis, V-3000
Catalog # 77674-916
Supplier:  CHEMetrics
New Product
undefined
Multi-Analyte Photometer for Water Quality Analysis, V-3000
Catalog # 77674-916
Supplier:  CHEMetrics
Supplier Number:  V-3000
New Product
Specifications
  • Description:
    V-3000 Multi-analyte photometer for water quality analysis
Specifications
About this item
CHEMetrics® handheld, portable V-3000 Multi-Analyte Water Photometer offers cutting edge technology for applications in the field and in the laboratory.
  • Field portable Rechargeable battery and universal power pack (Power LabStation required) Waterproof Web-based methods update Data logging storage for 100 data sets Language selections: English, German, French, Spanish
Intuitive and easy to use, the menu guides the analyst through all measuring tasks and is pre-programmed with over 40 calibrations using CHEMetrics Vacu-vials ampoules and MDL+ test kits.
Ordering information: Test kits sold separately.
Delivery information: The V-3000 model comes in a cardboard box and includes the multi-analyte water analysis photometer, 13 mm adapter and light shield, two 28 mm vials, four AA batteries, PC interface cable and operator's manual.