ContactUsLinkComponent
Ozone Single Analyte Photometer Kit
Ozone Single Analyte Photometer Kit
Catalog # 77674-706
Supplier:  CHEMetrics
New Product
undefined
Ozone Single Analyte Photometer Kit
Catalog # 77674-706
Supplier:  CHEMetrics
Supplier Number:  I-2023
New Product
Specifications
  • Description:
    Ozone single analyte photometer kit 0 - 0.25 ppm
Specifications
About this item
SAM (Single-Analyte Photometer) ozone kit.
  • Range 0 to 0.25 ppm Method: Indigo
Note: Both 77674-846 and 77674-844 Ozone Vacu-vials kits can be used with the 77674-706 SAM.
Delivery information: Kit comes in a plastic case and contains everything needed to perform 30 tests: Vacu-vials kit, SAM Photometer, four AAA batteries, screwdriver, and instructions.