Human Recombinant FGF-8a (from E. coli)
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Documents
TDS RUO Growth Factors Cytokines.pdf
Datasheet
- Catalog No:
- 77870-789
- 77870-842
- Protein/peptide name:
- FGF-8a
- FGF-8a
- Protein synonyms:
- Fibroblast Growth Factor-8a
- Fibroblast Growth Factor-8a
- Protein/peptide type:
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Protease-free:
- N
- N
- Animal-free:
- Y
- Y
- Nuclease-free:
- N
- N
- Carrier-free:
- Y
- Y
- Endotoxin-free:
- Y
- Y
- Storage conditions:
- Ambient
- Ambient
- Shipping temperature:
- Ambient
- Ambient
Product Family Options
- Pack typeSize
- : CYK-0025-0027
Specifications
Cat. No.77870-789Protein/peptide nameFGF-8aProtein synonymsFibroblast Growth Factor-8aProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - : CYK-0100-0027
Specifications
Cat. No.77870-842Protein/peptide nameFGF-8aProtein synonymsFibroblast Growth Factor-8aProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



