Human Recombinant TGF-β1 PLUS (from E. coli)
Supplier: Sartorius
About this item
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
3 Options Available Below
Product Details & Documents
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Documents
TDS RUO Growth Factors Cytokines.pdf
Datasheet
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77870-825
- 77870-814
- 77870-773
- Protein/peptide name:
- TGF-β1 PLUS
- TGF-β1 PLUS
- TGF-β1 PLUS
- Protein synonyms:
- Transforming Growth Factor-β1 PLUS
- Transforming Growth Factor-β1 PLUS
- Transforming Growth Factor-β1 PLUS
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Protease-free:
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Endotoxin-free:
- Y
- Y
- Y
- Storage conditions:
- Ambient
- Ambient
- Ambient
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: CYK-0100-0010Specifications:
More
Specifications
Cat. No.77870-825Protein/peptide nameTGF-β1 PLUSProtein synonymsTransforming Growth Factor-β1 PLUSProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0050-0010Specifications:
More
Specifications
Cat. No.77870-814Protein/peptide nameTGF-β1 PLUSProtein synonymsTransforming Growth Factor-β1 PLUSProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0025-0010Specifications:
More
Specifications
Cat. No.77870-773Protein/peptide nameTGF-β1 PLUSProtein synonymsTransforming Growth Factor-β1 PLUSProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient



