Human Recombinant BDNF (from E. coli)
Supplier: Sartorius
About this item
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
4 Options Available Below
Product Details & Documents
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Documents
TDS RUO Growth Factors Cytokines.pdf
Datasheet
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77870-766
- 77870-871
- 77870-823
- 77870-771
- Protein/peptide name:
- BDNF
- BDNF
- BDNF
- BDNF
- Protein synonyms:
- Brain Derived Neurotrophic Factor
- Brain Derived Neurotrophic Factor
- Brain Derived Neurotrophic Factor
- Brain Derived Neurotrophic Factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- Y
- Y
- Y
- Y
- Storage conditions:
- Ambient
- Ambient
- Ambient
- Ambient
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: CYK-0010-0008Specifications:
More
Specifications
Cat. No.77870-766Protein/peptide nameBDNFProtein synonymsBrain Derived Neurotrophic FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0500-0008Specifications:
More
Specifications
Cat. No.77870-871Protein/peptide nameBDNFProtein synonymsBrain Derived Neurotrophic FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0100-0008Specifications:
More
Specifications
Cat. No.77870-823Protein/peptide nameBDNFProtein synonymsBrain Derived Neurotrophic FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0025-0008Specifications:
More
Specifications
Cat. No.77870-771Protein/peptide nameBDNFProtein synonymsBrain Derived Neurotrophic FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



