Human Recombinant R-SP1 (from E. coli)
Supplier: Sartorius
About this item
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
3 Options Available Below
Product Details & Documents
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77870-782
- 77870-876
- 77870-834
- Protein/peptide name:
- R-SP1
- R-SP1
- R-SP1
- Protein synonyms:
- R-Spondin 1
- R-Spondin 1
- R-Spondin 1
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Protease-free:
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Endotoxin-free:
- Y
- Y
- Y
- Storage conditions:
- Ambient
- Ambient
- Ambient
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: CYK-0025-0019Specifications:
More
Specifications
Cat. No.77870-782Protein/peptide nameR-SP1Protein synonymsR-Spondin 1Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0500-0019Specifications:
More
Specifications
Cat. No.77870-876Protein/peptide nameR-SP1Protein synonymsR-Spondin 1Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0100-0019Specifications:
More
Specifications
Cat. No.77870-834Protein/peptide nameR-SP1Protein synonymsR-Spondin 1Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



