SafeTouch® Connect™ Latex Medical Examination Gloves
: Medicom
New Product
Prevents transmission of a wide variety of diseases.
Versatile, low protein content latex glove
- Fully textured for optimal grip
- Comfort and tactile sensitivity
Gloves act as a barrier to prevent contamination between patient and caregiver.
:
100 gloves per box, 10 boxes per case (500 pairs).
:
Latex allergies.
- Catalog No:
- CA77995-664
- CA77995-668
- CA77995-650
- CA77995-658
- CA77995-656
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 3.15 mil
- 3.15 mil
- 3.15 mil
- 3.15 mil
- 3.15 mil
- Palm thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- ASTM No.:
- ASTM D6124, ASTM D5712
- ASTM D6124, ASTM D5712
- ASTM D6124, ASTM D5712
- ASTM D6124, ASTM D5712
- ASTM D6124, ASTM D5712
Frequently Bought Together