Certified pH Buffer Solution, pH 7 at 20°C, Coloured (Yellow), with Fungicide, Paragon Scientific
Catalog # 77975-899
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Certified pH Buffer Solution, pH 7 at 20°C, Coloured (Yellow), with Fungicide
- Size:500 ml
- Certification:ISO 17034