Orders LinkContactUsLinkComponent
Mega Molecules Geologic Structures in Action Kit
Geologic structures in action kit, MH/HS, 3, 30
  470359-060
New Product
 :  GC10
Mega Molecules Geologic Structures in Action Kit
  470359-060
New Product
Mega Molecules Geologic Structures in Action Kit
Mega Molecules Geologic Structures in Action Kit
Geologic structures in action kit, MH/HS, 3, 30
$63.12
Each
A hands-on interactive model to teach tectonics, designed for the entire class.
  • Simulates real geologic structures: anticlines, synclines, and three types of faults: normal, reverse, and strike-slip
  • Made from soft, durable, non-toxic EVA foam (free from BPA, latex, and formaldehyde)
  • Supports NGSS MS-ESS2-2 for Earth science education
  • Ideal for repeated classroom use due to the material's resilience and reusability
The Geologic structures in action kit brings earth science to life through interactive modeling of rock deformation and tectonic activity. Students use the fold model to explore how layers of rock bend to form anticlines and synclines.  Students  physically manipulate the realistic, color-coded fault models to simulate the three major types of faults. This kit helps students construct evidence-based explanations of how earth’s surface changes over time. Durable and safe for classroom use, the kit is ideal for collaborative learning and long-term instruction.
 : Includes ten model packages (each with fold and fault models) for hands-on learning.
  • Description:
    Geologic structures in action kit
  • No. of lab groups:
    3
  • No. of students:
    30
  • Grade:
    MH/HS