Kidder Dancing Automata STEM Build Kit
Catalog # 470358-036
Supplier: KIDDER MFG. CO. LTD
New Product
Specifications
- Description:Dancing automata STEM build kit
Specifications
About this item
Construct a dancing automaton to explore cams and mechanical motion.
- Encourages hands-on STEM learning Demonstrates cam mechanisms converting rotational to linear motion Suitable for various educational levels Includes online teacher resource guide
The Automata Dancing STEM Build Kit offers students an engaging hands-on experience to explore the fundamentals of mechanical motion and cam mechanisms. By assembling a functional dancing automaton, learners gain insight into how rotational motion can be converted into linear motion using cams. This activity not only reinforces theoretical knowledge but also enhances problem-solving skills, making it an excellent addition to STEM education programs across various grade levels.
Application: Engineering, Mechanics