Kidder Elevator STEM Build Kit
Elevator STEM build kit
Catalog # 470358-026
Supplier: KIDDER MFG. CO. LTD
Supplier #: 80-50-W025
Jump to:
Product Details & Documents
Construct a motorized elevator to explore simple machines and vertical transport systems.
- Encourages hands-on STEM learning Demonstrates pulley systems and gear mechanisms Includes online teacher resource guide
The Elevator STEM Build Kit offers students a practical introduction to mechanical systems and simple machines. By assembling a functional elevator model powered by a DC motor and gears, learners can observe the principles of vertical transportation, including the roles of pulleys and guide tracks. This hands-on activity reinforces concepts such as force, motion, and mechanical advantage, making it an excellent addition to STEM education programs.
Application: Engineering; Mechanics
Documents
Specifications
- Description:Elevator STEM build kit