ContactUsLinkComponent
Four-Way Digital Soil Analyzer
Four-Way Digital Soil Analyzer
Catalog # 470358-646
Supplier:  Luster Leaf
New Product
undefined
Four-Way Digital Soil Analyzer
Catalog # 470358-646
Supplier:  Luster Leaf
Supplier Number:  1885
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Four-way digital soil analyzer
Specifications
About this item
Determines proper moisture, pH, light, and temperature in soil samples.
  • Large and easy to read digital display
  • Probe length 5"
  • Four function switch
Ensure healthy vegetative growth with accurate, calibrated readings. This device measures light levels adjusted for optimal theoretical plant growth across all light sources, including variations in natural daylight. Calibrated to the USDA mean solar radiation table, it provides reliable, research-based light data.
Also features soil temperature measurement in both fahrenheit and celsius, with an easy-to-reference guide showing ideal germination and growth temperature ranges for a wide variety of plants.