ContactUsLinkComponent
LaMotte SpinDisks® Reagent Cartridges for Fresh Water Testing
LaMotte SpinDisks® Reagent Cartridges for Fresh Water Testing
Catalog # 470358-630
Supplier:  LaMotte
New Product
LaMotte SpinDisks® Reagent Cartridges for Fresh Water Testing
Catalog # 470358-630
Supplier:  LaMotte
Supplier Number:  4353-H
New Product
Specifications
  • Description:
    Aquaculture fresh water disks (pack of 50)
  • Environmentally Preferable:
Specifications
About this item
Test multiple freshwater parameters in minutes with pre-measured, no-mess reagent disks.
  • Pre-measured reagents for consistent freshwater testing
  • Designed specifically for freshwater aquaculture applications
  • No mixing or measuring - just fill the disk and test
  • Compatible with LaMotte Spin Touch® FX photometer
  • Run multiple tests from a single water sample
  • Sealed design improves safety and reduces waste
  • Speeds up classroom or field testing for teachers and students
  • Supports hands-on science and STEM education
The FX Fresh Water Series SpinDisks® are designed to simplify water testing in educational and field environments. Created for use with the LaMotte Spin Touch® FX photometer, these sealed reagent disks provide fast, reliable freshwater analysis with no measuring, mixing, or cleanup required.
Each SpinDisk® contains all the reagents needed to test multiple parameters from a single water sample. Simply fill the disk, place it in the Spin Touch FX, and receive accurate results in just 2 minutes. This all-in-one approach saves time and reduces the risk of user error, making it ideal for students and teachers alike.
These disks are perfect for hands-on activities in environmental science, biology, chemistry, or general STEM programs. Teachers can confidently bring water quality testing into the classroom or field without the mess or safety concerns of traditional testing methods.
FX Fresh Water Series SpinDisks® support inquiry-based learning and data-driven science lessons by giving students a real-world tool for exploring aquatic ecosystems and water chemistry.
Ordering information:
LaMotte Spin Touch® FX photometer not included (sold separately).