HOBO® State Data Logger
Catalog # 470323-540
Supplier: ONSET COMPUTER CORPORATION SE
Specifications
- Description:HOBO® state data logger
Specifications
About this item
Records state changes, electronic pulses, and mechanical or electrical contact closures from external sensing devices.
One logger with four recording modes
- Internal reed switch can be used to determine when a door or window is open or closed
- Compatible with a wide range of external sensors
- Ships with 2.5 mm input cable
- Compatible with HOBOware and HOBOware Pro software for logger setup and graphing/analysis
Ideal for monitoring energy consumption, mechanical equipment operation, and water and gas flow. 1 Hz maximum state, event, runtime frequency.