ContactUsLinkComponent
Eisco® Metal Flywheel
Catalogue # 470344-500
Fournisseur:  Wards
undefined
Eisco® Metal Flywheel
Catalogue # 470344-500
Fournisseur:  Wards
Supplier Number:  PH0313
Spécifications
  • Weight:
    8.62 kg (19 lbs.)
  • Diameter:
    190.5 mm (7.5")
Spécifications
À propos de cet article
Balanced flywheel for exploring inertia and rotational dynamics.
  • Ball-bearing supported horizontal shaft
  • Bracket includes fixed pointer for observation
  • Can be wall-mounted using four integrated holes
Ideal for physics labs, this flywheel enables in-depth studies of rotational energy, kinetic energy, and inertia. The wheel is carefully balanced to ensure smooth motion and accurate results during demonstrations. While the apparatus is ready for use out of the box, additional weights can be introduced for extended experimentation (not included).