ContactUsLinkComponent
VWR® Mini Digital Incubators, Heating, Heating and Cooling, CO₂
VWR® Mini Digital Incubators, Heating, Heating and Cooling, CO₂
Supplier:  VWR International
New Product
Certificates
undefined
VWR® Mini Digital Incubators, Heating, Heating and Cooling, CO₂
Supplier:  VWR International
New Product
Specifications
  • Catalog No:
  • 77553-336
  • 77553-340
  • 77553-338
  • Ext. W×D×H:
  • 26.1×23.6×32.5 cm (10.3×9.3×12.8")
  • 26.1×23.6×32.5 cm (10.3×9.3×12.8")
  • 26.1×23.6×32.5 cm (10.3×9.3×12.8")
  • Sensor:
  • Dual beam infrared
  • Platform dimensions:
  • 15×20.3 cm (6×8")
  • 15×20.3 cm (6×8")
  • 15×20.3 cm (6×8")