The pure² Aseptic Isolator is an ideal solution for safe and efficient aseptic processing. Designed for a wide variety of critical aseptic processes, the pure² pairs the highest quality materials and technologies for unmatched protection of products and users.
Maintains unidirectional Grade A (ISO Class 5) conditions
- Smooth and easy to clean 316L stainless steel interior
- Selectable positive or negative pressure operation
- Large and fast airlocks to improve isolator productivity
- High performance nanox catalytic converters for short decontamination cycles
- Fully integrated, fast and safe H₂O₂ decontamination cycles with skanfog technology within each chamber
- HEPA filtered supply air (H14) and exhaust air (dual H14)
- DIN 12980 compliant for cytotoxic aseptic-toxic applications
The pure² Aseptic Isolator is a high-performance aseptic isolator designed for critical process applications requiring Grade A (ISO Class 5) conditions. This isolator ensures sterile, contamination-free handling through advanced HEPA filtration, H₂O₂ decontamination, and precise environmental control, making it ideal for applications that demand the highest level of product and user protection.
The pure² is engineered for aseptic processing, sterility testing, and contamination-sensitive workflows. It is commonly used in pharmaceutical applications where product protection is critical. With its modular design, customizable features, and fast decontamination cycles, the pure² isolator enhances productivity, flexibility, and operational efficiency, making it a superior choice for high-precision, contamination-controlled environments.
Available in 2 and 4 glove configurations, the modular space-saving design is flexible and customizable, with a diverse range of customizations and integrations such as pass-throughs and environmental monitoring.
IQ/OQ documentation package includes testing procedures and documentation for regulatory compliance. Onsite microbiological qualification H₂O₂ decontamination cycles with BIs, aeration confirmation, residual detection, and full reporting.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.