ContactUsLinkComponent
Ritmed® AssureWear™ VersaGown® Flexneck™ Technology Chemotherapy Impervious Isolation Gowns
Ritmed® AssureWear™ VersaGown® Flexneck™ Technology Chemotherapy Impervious Isolation Gowns
Supplier:  Medicom
New Product
Certificates
undefined
Ritmed® AssureWear™ VersaGown® Flexneck™ Technology Chemotherapy Impervious Isolation Gowns
Supplier:  Medicom
New Product
Specifications
  • Catalog No:
  • CA76545-396
  • CA76544-854
  • CA76544-856
  • CA76544-858
  • CA76544-860
  • CA76544-862
  • CA76544-864
  • 76545-396
  • 76544-860
  • 76544-858
  • 76544-864
  • 76544-854
  • 76544-856
  • Closure type:
  • Back fastening
  • Back fastening
  • Pockets:
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Seam type:
  • Ultrasonically welded
  • Garment application:
  • Laboratory
  • Medical