Orders LinkContactUsLinkComponent
Sorenson™ Universal TipStation Pipette Tips
Sorenson™ Universal TipStation Pipette Tips
Supplier:   Sorenson Bioscience
Sorenson™ Universal TipStation Pipette Tips
Certificates
Sorenson™ Universal TipStation Pipette Tips
Sorenson™ Universal TipStation Pipette Tips
3 Options Available Below
Product Details & Documents
Universal Fit TipStation pipette tips are available sterile or non-sterile in 20 and 200 µl, and are a perfect platform for multi-channel pipetting.
  • Ideal platform for multi-channel pipettors or high throughput laboratories
  • Can position lid to expose 96 or 192 tips
  • Certified RNase/DNase free, human DNA free, and non-pyrogenic
  • Wide compatibility across popular pipettors
Certifications: ISO 13485, ISO 14001.
Documents
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    Length
    Tip color
    For use with
    Packed
    Availability
    Price
  • Universal fit, extended length
    0.1 - 10 µl
    -
    45.34 mm
    Clear
    Universal pipettors
    5 layers, 192 each
    Discontinued
    Supplier Number: -
  • Universal fit, extended length
    0.1 - 10 µl
    +
    45.34 mm
    Clear
    Universal pipettors
    5 layers, 192 each
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 960
    Pack of 960 - $124.84
    $124.84
    New Customer Promotional Price
    Log in for availability
    Case of 4800
    Case of 4800 - $624.19
    $624.19
    New Customer Promotional Price
    Pack of 960
       $124.84
    Case of 4800
       $624.19
    Supplier Number: 27790
el 4
Product Information
  • Tip style
    Volume
    Sterile
    Length
    Tip color
    For use with
    Packed
    Availability
    Price
  • Universal fit
    1 - 200 µl
    51.44 mm
    Clear
    Universal pipettors
    5 layers, 192 each
    Log in for availability
    Log in for availability
    Case of 4,800
    Case of 4,800 - $181.33
    $181.33
    New Customer Promotional Price
    Case of 4,800
       $181.33
    Supplier Number: 27760
  • Universal fit
    1 - 200 µl
    51.44 mm
    Clear
    Universal pipettors
    5 layers, 192 each
    Log in for availability
    Log in for availability
    Case of 4,800
    Case of 4,800 - $216.63
    $216.63
    New Customer Promotional Price
    Case of 4,800
       $216.63
    Supplier Number: 27770