CiDehol® 70 70% IPA Solution for Endotoxin
Catalog # 10810-848
Supplier: Decon Labs
Specifications
- Description:CiDehol® 70 70% Isopropyl alcohol
- Packaging:Plastic drum
Specifications
About this item
70% Isopropyl alcohol supplied in a 55 gal. plastic drum, tested for endotoxins.
IPA solution evaporates away completely so no rinse is required
- Filtered to 0.2 µm
- Use to clean surfaces in microbiology labs and tissue culture labs
CiDehol® 70 is a 70% (v/v) Isopropyl alcohol solution filtered to 0.2 microns. CiDehol® 70 can be used to clean surfaces in microbiology labs and tissue culture labs. It is also effective in cleaning surfaces and process equipment in pharmaceutical, biotechnology, and medical device facilities.