Order Entry
Canada
Orders LinkContactUsLinkComponent
Ward's® Agate

Ward's® Agate

Agate, bulk pack, 1 kg, Size will vary
  470357-982
 :   Avantor
Ward's® Agate
  470357-982
 :  Avantor
Ward's® Agate
Ward's® Agate

Agate, bulk pack, 1 kg, Size will vary

 
$36.32
   
Each
 

Clear to yellow translucent.

  • Oregon

The number and size of pieces will vary, but typically average between 1×2" to 2×3".

 
  • Description:
    Agate, bulk pack, 1 kg
  • Specimen dimensions:
    Size will vary
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.