Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CXCL12 (SDF-1a) (from E. coli)
Human Recombinant CXCL12 (SDF-1a) (from E. coli)
Certificates
undefined
Human Recombinant CXCL12 (SDF-1a) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Chemokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Avoid repeated freeze-thaw cycles 12 months from date of receipt, –20...–70 °C as supplied. 1 month, 2...8 °C under sterile conditions after reconstitution. 3 months, –20...–70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.01 EU per 1 μg of the protein by the LAL method
  • Biological activity:
    EC50 = 0.62 nM determined by Calcium Flux with human CXCR4 in U937 cells EC50 = 0.17nM determined by Migration with U937 cells
  • Gene ID:
    Accession # P48061-2 (22-89)
  • Reconstitution instructions:
    Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
  • Endotoxin-free:
    Y
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Protein synonyms:
    IRH|PBSF|SDF1|TLSF|TPAR1|SCYB12
  • Protein/peptide name:
    CXCL12 (SDF-1a)
  • Purity:
    >97%
  • Molecular weight:
    7.96 kDa
  • Sequence:
    KPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNK
  • Endotoxin level:
    Very low
  • Formulation:
    Lyophilized pellet
  • Shipping temperature:
    RT
  • Tested applications:
    Migration assay; Calcium Flux

Specifications

Frequently Bought Together