Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CXCL8 (IL-8) (from E. coli), Biotinylated
Human Recombinant CXCL8 (IL-8) (from E. coli), Biotinylated
Certificates
undefined
Human Recombinant CXCL8 (IL-8) (from E. coli), Biotinylated

Specifications

  • Conjugation:
    Biotin
  • Protein Function:
    Chemokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Avoid repeated freeze-thaw cycles. 12 months from date of receipt, –20... –70 °C as supplied. ChemoTactics suggests using immediately after reconstitution. 1 month, –20...–70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.01 EU per 1 μg of the protein by the LAL method
  • Biological activity:
    EC50 = 0.5 - 1.0 nM determined by Migration Assay in cells expressing recombinant CXCR1
  • Gene ID:
    Accession # P10145 (28-99)
  • Reconstitution instructions:
    Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
  • Endotoxin-free:
    Y
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Protein synonyms:
    IL8|NAF|GCP1|LECT|LUCT|NAP1|GCP-1|LYNAP|MDNCF|MONAP|NAP-1|SCYB8
  • Protein/peptide name:
    CXCL8 (IL-8)
  • Purity:
    >97%
  • Molecular weight:
    10.80 kDa
  • Endotoxin level:
    Very low
  • Formulation:
    Lyophilized pellet
  • Shipping temperature:
    RT
  • Tested applications:
    Migration assay

Specifications

Frequently Bought Together