Human Recombinant CCL7 (MCP-3) (from E. coli), Biotinylated
Supplier: CHEMOTACTICS, INC MS
Certificates
About this item
Functional biotin labeled human CCL7 chemokine with very low endotoxin levels.
- Extremely low endotoxin levels (<0.01 EU per 1 μg) Functional activity tested and validated For use in various streptavidin applications: Flow cytometry, microscopy, and other imaging techniques
Monocyte chemotactic protein 3 (MCP3)(CCL7) is secreted by monocytes and certain tumer cell lines. It is a chemoattractant for monocytes and other leukocytes. MCP3 binds and signals through several cell surface receptors including CCR1, CCR2, CCR3, and appears to be an antagonist for CCR5.
Ordering information: Larger sizes available, please request a custom quote.
Delivery: Shipped lyophilized at room temperature.
Caution: For research use only, not for use in humans.
Specifications
- Conjugation:Biotin
- Protein Function:Chemokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Avoid repeated freeze-thaw cycles. 12 months from date of receipt, –20... –70 °C as supplied. ChemoTactics suggests using immediately after reconstitution. 1 month, –20...–70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.01 EU per 1 μg of the protein by the LAL method
- Biological activity:EC50 = 1.4 - 2.6 nM determined by migration assay with cells expressing recombinant CCR2
- Gene ID:Accession # P80098 (24-99)
- Reconstitution instructions:Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
- Endotoxin-free:Y
- Carrier-free:Y
- Protease-free:Y
- Protein synonyms:FIC|MARC|MCP-3|MCP3|NC28|SCYA6|SCYA7
- Protein/peptide name:CCL7 (MCP-3)
- Purity:>97%
- Molecular weight:11.38 kDa
- Endotoxin level:Very low
- Formulation:Lyophilized pellet
- Shipping temperature:RT
- Tested applications:Migration assay
Specifications
Frequently Bought Together