Human Recombinant CCL19 (MIP-3beta) (from E. coli), Biotinylated
Supplier: CHEMOTACTICS, INC MS
Certificates
About this item
Functional biotin labeled human CCL19 chemokine with very low endotoxin levels.
- Extremely low endotoxin levels (<0.01 EU per 1 μg) Functional activity tested and validated For use in various streptavidin applications: Flow cytometry, microscopy, and other imaging techniques
Macrophage inflammatory protein-3-beta (MIP3β) (CCL19), also known as EBI1 ligand chemokine (ELC), directs chemotaxis of dendritic cells, and certain B- and T- lymphocytes, but not monocytes or granulocytes. It is constitutively expressed in thymus and lymph nodes and binds specifically to target cells expressing the receptor CCR7. Being a homeostatic chemokine, its primary physiological role is considered to be in the normal recirculation and homing of lymphocyte. However, MIP3β could also be proinfammatory, and is implicated in the post-HIV infection responses.
Ordering information: Larger sizes available, please request a custom quote.
Delivery: Shipped lyophilized at room temperature.
Caution: For research use only, not for use in humans.
Specifications
- Conjugation:Biotin
- Protein Function:Chemokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Avoid repeated freeze-thaw cycles. 12 months from date of receipt, –20... –70 °C as supplied. ChemoTactics suggests using immediately after reconstitution. 1 month, –20...–70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.01 EU per 1 μg of the protein by the LAL method
- Biological activity:EC50 = 15 - 30 nM determined by migration assay with cells expressing recombinant CCR7
- Gene ID:Accession Q99731 (22-98)
- Reconstitution instructions:Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
- Endotoxin-free:Y
- Carrier-free:Y
- Protease-free:Y
- Protein synonyms:CKb11|ELC|MIP-3b|MIP3B|SCYA19
- Protein/peptide name:CCL19 (MIP-3beta)
- Purity:>97%
- Molecular weight:11.22 kDa
- Endotoxin level:Very low
- Formulation:Lyophilized pellet
- Shipping temperature:RT
- Tested applications:Migration assay
Specifications
Frequently Bought Together