Order Entry
Canada
ContactUsLinkComponent
ZymoPURE II™ Plasmid Gigaprep Kit
ZymoPURE II™ Plasmid Gigaprep Kit
Catalog # 77001-472
Supplier:  Zymo Research
CAS Number:  
undefined
ZymoPURE II™ Plasmid Gigaprep Kit
Catalog # 77001-472
Supplier:  Zymo Research
Supplier Number:  D4204
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Giga
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    ZymoPURE™ II Plasmid Gigaprep Kit
  • Size:
    5 preps
  • Sample size:
    ≤2.5 l
  • Description:
    ZymoPURE™ II Plasmid Gigaprep Kit
  • Yield:
    ≤10 mg
  • Cat. no.:
    77001-472

Specifications

About this item

Part of the ZymoPURE plasmid kits collection, the ZymoPURE II™ plasmid midiprep kits provides the fastest and simplest method available to efficiently isolate up to 1.2 of transfection grade plasmid DNA from E. coli.

  • Fastest: Simple 20 minute Midi/Maxi preps (50 min Giga)
  • Highest Yield: 6× more plasmid
  • Ultra-Pure: EndoZero™, vaccine grade and transfection ready

Utilizing a modified alkaline lysis in conjunction with a binding system, the ZymoPURE II midiprep kits can process up to 50 ml of culture in less than 18 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is rapidly bound onto a column with either a vacuum or a centrifuge instead of a slow gravity flow column. Additionally, there is no alcohol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 6 µg/µl), endotoxin-free and transfection-ready. As an added convenience, the ZymoPURE II™ plasmid midiprep kits contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.