Order Entry
Canada
ContactUsLinkComponent
ZymoPURE II™ Plasmid Maxiprep Kits
ZymoPURE II™ Plasmid Maxiprep Kits
Supplier:  Zymo Research
Certificates
undefined
ZymoPURE II™ Plasmid Maxiprep Kits
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample size:
    ≤150 ml
  • Description:
    ZymoPURE™ II Plasmid Maxiprep Kit
  • Yield:
    ≤1.2 mg

Specifications

Frequently Bought Together