About this item
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down.
Main post diameter: 14 mm
Sub post diameter: 12 mm
Base: 50W×58D×55H mm (2.0W×2.3D×2.2” H)
Vertical holding power: 176 lb.
Clamp hole: Adjustable from 4.5 to 6.5 mm
M10: This magnetic base is similar to M1 but with a 12 mm Ø sub pole that fits the 12 mm clamp supplied with M3301, DC3001, MD4 and MMJ manipulators. These are an ideal manipulator stands.
M10L: This magnetic base is the same as the M10 (12 mm Ø sub pole), but it is equipped with a taller (14") vertical main pole. These are an ideal manipulator stands.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.