Human/Mouse Recombinant NT-3 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Neurotrophin-3 (NT-3) is an important member of the nerve growth factor (NGF) family of proteins. NT-3 promotes the growth, survival, and differentiation of neurons and synapses in the peripheral and central nervous systems. The receptor tyrosine kinase TrkC exclusively binds in high-affinity to NT-3. NT-3 also signals through the receptor tyrosine kinase TrkB, and through the low affinity nerve growth factor receptor (LNGFR).
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage conditions:At or below −20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6); ED₅₀ ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
- Gene ID:P20783/P20181
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Neurotrophin 3|Nerve growth factor 2 (NGF-2)|HGNF|NT3|Neurotrophic Factor
- Protein/peptide name:NT-3
- Purity:≥95%
- Molecular weight:13.8/27.5 kDa
- Sequence:MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together