Order Entry
Canada
ContactUsLinkComponent
 
Human/Mouse Recombinant NT-3 (from E. coli)
New Product
 
undefined
Human/Mouse Recombinant NT-3 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6); ED₅₀ ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
  • Gene ID:
    P20783/P20181
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Neurotrophin 3|Nerve growth factor 2 (NGF-2)|HGNF|NT3|Neurotrophic Factor
  • Protein/peptide name:
    NT-3
  • Purity:
    ≥95%
  • Molecular weight:
    13.8/27.5 kDa
  • Sequence:
    MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together