Order Entry
Canada
ContactUsLinkComponent
 
Dog Recombinant GM-CSF (from E. coli)
New Product
 
undefined
Dog Recombinant GM-CSF (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Dog
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation; ED₅₀ ≤15 ng/ml; Bioactivity ≥6.7×10⁴ units/mg
  • Gene ID:
    P26891
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    T cell growth factor|TCGF|Aldesleukin
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥95%
  • Molecular weight:
    15.3 kDa
  • Sequence:
    MAPTSSSTKN TKKQLEPLLL DLQLLLKEVK NYENADLSRM LTFKFYMPKQ ATELKHLQCL VEELKALEGV LNLGQSKNSD SANIKESMNN INVTVLELKG SETSFKCEYD DETVTAVEFL NKWITFCQSI YSTLT
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together