Mouse Recombinant MIP-2 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Macrophage Inflammatory Protein 2 (MIP-2), also known as CXCL2, is a small cytokine that is secreted by monocytes and neutrophils at sites of inflammation. MIP-2 functions through the chemokine receptor CXCR2 to act as a chemotactic agent for leukocytes and hematopoietic cells.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P10889
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL2
- Protein/peptide name:MIP-2
- Purity:≥95%
- Molecular weight:7.9 kDa
- Sequence:AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together