Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IL-19 (from E. coli)
Mouse Recombinant IL-19 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Mouse Recombinant IL-19 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q8CJ70
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein/peptide name:
    IL-19
  • Purity:
    ≥95%
  • Molecular weight:
    17.7 kDa
  • Sequence:
    MLRRCLISVDMRLIEKSFHEIKRAMQTKDTFKNVTILSLENLRSIKPGDVCCMTNNLLTFYRDRVFQDHQERSLEVLRRISSIANSFLCVQKSLERCQVHRQCNCSQEATNATRIIHDNYNQLEVSSAALKSLGELNILLAWIDRNHLETPAA
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together