Order Entry
Canada
ContactUsLinkComponent
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
    Rat
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Alkaline phosphatase activity induced in ATDC-5 cells; ED₅₀ ≤250 ng/ml; Bioactivity ≥4.0×10³ units/mg
  • Gene ID:
    P12643/P21274/P49001
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    BMP-2A
  • Protein/peptide name:
    BMP-2 (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    13.0/26.1 kDa
  • Sequence:
    MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together