Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant Follistatin (from E. coli)
Mouse Recombinant Follistatin (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Mouse Recombinant Follistatin (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤5
  • Biological activity:
    No biological activity data is available at this time
  • Gene ID:
    P47931
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    FS|activin-binding protein|FSH-suppressing protein (FSP)
  • Protein/peptide name:
    Follistatin
  • Purity:
    ≥95%
  • Molecular weight:
    31.6 kDa
  • Sequence:
    MGNCWLRQAK NGRCQVLYKT ELSKEECCST GRLSTSWTEE DVNDNTLFKW MIFNGGAPNC IPCKETCENV DCGPGKKCRM NKKNKPRCVC APDCSNITWK GPVCGLDGKT YRNECALLKA RCKEQPELEV QYQGKCKKTC RDVFCPGSST CVVDQTNNAY CVTCNRICPE PSSSEQYLCG NDGVTYSSAC HLRKATCLLG RSIGLAYEGK CIKAKSCEDI QCGGGKKCLW DSKVGRGRCS LCDELCPDSK SDEPVCASDN ATYASECAMK EAACSSGVLL EVKHSGSCN
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together