Mouse Recombinant SCF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together