Human Recombinant CD40-Ligand (from E. coli)
New Product
CD40-Ligand, or CD154, is a membrane glycoprotein and differentiation antigen that is expressed on the surface of T cells, monocytes, basophils, eosinophils, platelets, dendritic cells, and endothelial cells. CD40-Ligand binds and activates the CD40 receptor on antigen-presenting cells. CD40-Ligand stimulates B cell proliferation, immunoglobulin class switching, and antibody secretion.
High quality
- Low endotoxin
- Affordable
CD40-Ligand also induces cytokine production and tumoricidal activity in peripheral blood monocytes. CD40-Ligand is a co-stimulator of activated T cell proliferation and inflammatory protein production.
:
For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Activity of HEK-blue™ CD40L cells; ED₅₀ ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
- Gene ID:P29965
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:TNFSF5|TRAP|CD154|gp39|T-BAM
- Protein/peptide name:CD-40-Ligand (AF)
- Purity:≥95%
- Molecular weight:16.3 kDa
- Sequence:MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together