Human Recombinant CTGF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Connective tissue growth factor (CTGF) is a mitogen that is secreted by vascular endothelial cells in response to basic fibroblast growth factor (bFGF) or vascular endothelial growth factor (VEGF). CTGF promotes cell growth, migration, adhesion, and survival of endothelial cells. CTGF is also important during osteogenesis, chondrogenesis, and skeletogenesis. CTGF has an insulin-like growth factor binding protein (IGFBP) domain, a thrombospondin type 1 repeat (TSR) domain, and a C-terminal cysteine knot motif.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: HUVEC cell proliferation; ED50 ≤2000 ng/ml; Bioactivity ≥5.0×10² units/mg
- Gene ID:P29279
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCN2|HCS24
- Protein/peptide name:CTGF
- Purity:≥95%
- Molecular weight:11.2 kDa
- Sequence:MGKKCIRTPK ISKPIKFELS GCTSMKTYRA KFCGVCTDGR CCTPHRTTTL PVEFKCPDGE VMKKNMMFIK TCACHYNCPG DNDIFESLYY RKMYGDMA
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together