About this item
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources. The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic. Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.
High quality
- Low endotoxin
- Affordable
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R-3T3 cell proliferation; ED₅₀ ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P09038
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:FGF2|HBGF-2|Prostatropin
- Protein/peptide name:FGF-Basic 147
- Purity:≥95%
- Molecular weight:16.5 kDa
- Sequence:#N/A
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay