Human Recombinant PLGF-3 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Placenta growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family and promotes endothelial cell growth and angiogenesis. PlGF is secreted as a homodimer and may also form a heterodimer with VEGF. PlGF is detected in the placenta, heart, lungs, thyroid, and adipose tissues. Circulating PlGF levels are correlated with colorectal and renal cancers, as well as atherosclerosis and ischemic heart disease. There are four alternatively spliced PlGF isoforms (PlGF-1, PlGF-2, PlGF-3, and PlGF-4), each with unique secretion patterns and heparin-binding affinities. PlGF-3 lacks a heparin-binding domain and signals through the VEGFR-1 receptor.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NA; ED50 ≤NA; Bioactivity ≥NA
- Gene ID:P49763
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:PLGF-3
- Purity:≥95%
- Molecular weight:22.9/45.8 kDa
- Sequence:MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRHSPGRQ SPDMPGDFRA DAPSFLPPRR SLPMLFRMEW GCALTGSQSA VWPSSPVPEE IPRMHPGRNG KKQQRKPLRE KMKPERCGDA VPRR
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together