Human Recombinant TNF-A (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS). TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P01375
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:TNFSF2|Cachectin|DIF|Necrosin|Cytotoxin|Cachexin|TNF
- Protein/peptide name:TNF-A
- Purity:≥95%
- Molecular weight:17.5 kDa
- Sequence:MVRSSSRTPS DKPVAHVVAN PQAEGQLQWL NRRANALLAN GVELRDNQLV VPSEGLYLIY SQVLFKGQGC PSTHVLLTHT ISRIAVSYQT KVNLLSAIKS PCQRETPEGA EAKPWYEPIY LGGVFQLEKG DRLSAEINRP DYLDFAESGQ VYFGIIAL
- Endotoxin level:Low
- Formulation:10 mM Sodium Phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together