Order Entry
Canada
ContactUsLinkComponent
Human Recombinant TPO (CHO) (from CHO cells)
Human Recombinant TPO (CHO) (from CHO cells)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant TPO (CHO) (from CHO cells)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: MO7e cell proliferation; ED₅₀ ≤15 ng/ml; Bioactivity ≥6.7×10⁴ units/mg
  • Gene ID:
    P40225
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Megakaryocyte Colony Stimulating Factor|c-MPL ligand|MGDF
  • Protein/peptide name:
    TPO
  • Purity:
    ≥90%
  • Molecular weight:
    36.3 kDa
  • Sequence:
    #N/A
  • Endotoxin level:
    Low
  • Formulation:
    5 mM Sodium phosphate, 2 mM L-Arginine, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together