Order Entry
Canada
Orders LinkContactUsLinkComponent
Proteometer-UFT Buffer Packets

Proteometer-UFT Buffer Packets

Proteometer-UFT Buffer Packets (3X)
Catalog # 77775-504
Supplier:   NOVILYTIC, LLC
Supplier #:  BUF-PUF-001
Proteometer-UFT Buffer Packets
Catalog # 77775-504
Supplier:  NOVILYTIC, LLC
CAS Number:  
Proteometer-UFT Buffer Packets
Proteometer-UFT Buffer Packets

Proteometer-UFT Buffer Packets (3X)

CAS Number:  
Price
$647.17
Log in for availability
Pack of 4
Product Details & Documents

3 sealed buffer packets specifically formulated for use with Proteometer-UFT kit.

  • Buffer packets available for reorder when the Proteometer-UFT MD Unit and/or Proteometer-UFT reagent are not required

Intended for use with the Proteometer-UFT kit.

Ordering information: Reorder individually as needed.

Packaging: Supplied in a custom box with instructions for use. Store at room temperature.

Specifications

Specifications

  • Description:
    Proteometer-UFT Buffer Packets (3X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.